Product Details

SNP ID
rs199628554
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:55369827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCAGGTCGGTTTCTTCATCGCTG[A/C]CGCGCTCATCTCCGCTCTCATCTGA
Phenotype
MIM: 613515
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ATG14 PubMed Links

Gene Details

Gene
ATG14
Gene Name
autophagy related 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014924.4 1011 Missense Mutation GGC,GTC G424V NP_055739.2
XM_011536563.2 1011 Missense Mutation GGC,GTC G311V XP_011534865.1

View Full Product Details