Product Details

SNP ID
rs201478064
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:66341169 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTACCATTTCCTAGGGTCTGACT[A/G]TTACTCAGCAGCTTTGCCTGCCTTC
Phenotype
MIM: 614183 MIM: 610716
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DIS3L PubMed Links

Gene Details

Gene
DIS3L
Gene Name
DIS3 like exosome 3'-5' exoribonuclease
There are no transcripts associated with this gene.

Gene
SCARNA14
Gene Name
small Cajal body-specific RNA 14
There are no transcripts associated with this gene.

Gene
TIPIN
Gene Name
TIMELESS interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289986.1 932 Intron NP_001276915.1
NM_017858.2 932 Intron NP_060328.2
XM_005254521.2 932 Silent Mutation AAC,AAT N221N XP_005254578.1
XM_005254522.4 932 Silent Mutation AAC,AAT N221N XP_005254579.1
XM_006720594.3 932 Silent Mutation AAC,AAT N221N XP_006720657.1
XM_011521743.1 932 Silent Mutation AAC,AAT N221N XP_011520045.1
XM_017022390.1 932 Silent Mutation AAC,AAT N221N XP_016877879.1
XM_017022391.1 932 Intron XP_016877880.1

View Full Product Details