Product Details

SNP ID
rs201968578
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:70832127 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGACCCCTGGGCAACCTCAGCAC[C/T]CCTACGGGTAGCCCATCTGCAGTCT
Phenotype
MIM: 611300
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
LARP6 PubMed Links

Gene Details

Gene
LARP6
Gene Name
La ribonucleoprotein domain family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286679.1 1313 Silent Mutation GGA,GGG G283G NP_001273608.1
NM_018357.3 1313 Silent Mutation GGA,GGG G467G NP_060827.2
NM_197958.2 1313 Intron NP_932062.1

View Full Product Details