Product Details

SNP ID
rs201336806
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:42725172 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCACTGGGCCTGGTGGAGGCTG[C/T]CCAGGCAGGCCTCTATCTCCATCCG
Phenotype
MIM: 607465 MIM: 614642
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDAN1 PubMed Links

Gene Details

Gene
CDAN1
Gene Name
codanin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138477.2 3587 Missense Mutation GAC,GGC D1177G NP_612486.2
XM_005254176.4 3587 Missense Mutation GAC,GGC D1178G XP_005254233.1
XM_011521270.2 3587 Missense Mutation GAC,GGC D1186G XP_011519572.1
XM_011521271.2 3587 Missense Mutation GAC,GGC D1185G XP_011519573.1
XM_011521274.2 3587 Missense Mutation GAC,GGC D841G XP_011519576.1
Gene
STARD9
Gene Name
StAR related lipid transfer domain containing 9
There are no transcripts associated with this gene.

View Full Product Details