Product Details

SNP ID
rs207475597
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:74749948 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGGCACTGTCAAGGATGAGCCA[A/G]CGCTACGGGGACGTCCTGCAGATCC
Phenotype
MIM: 124060
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYP1A2 PubMed Links

Gene Details

Gene
CYP1A2
Gene Name
cytochrome P450 family 1 subfamily A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000761.4 274 Silent Mutation CAA,CAG Q70Q NP_000752.2

View Full Product Details