Product Details

SNP ID
rs201237590
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:101656997 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGTACAACTGTGATCCAAGTCAA[C/T]GTCAGCCATAAATCCTTCTTCAAAA
Phenotype
MIM: 610014
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TARSL2 PubMed Links

Gene Details

Gene
TARSL2
Gene Name
threonyl-tRNA synthetase like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152334.2 1690 Missense Mutation ATT,GTT I729V NP_689547.2
XM_006720386.3 1690 Silent Mutation ACA,ACG T704T XP_006720449.1
XM_017021912.1 1690 Intron XP_016877401.1
XM_017021913.1 1690 Missense Mutation ATT,GTT I486V XP_016877402.1
Gene
TM2D3
Gene Name
TM2 domain containing 3
There are no transcripts associated with this gene.

View Full Product Details