Product Details

SNP ID
rs201567790
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:75904541 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCGCAGAGTATTGCGGACCCATC[A/G]GAGCGTAACCTGGATCTCCGCAGGC
Phenotype
MIM: 609096 MIM: 612501
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXO22 PubMed Links

Gene Details

Gene
FBXO22
Gene Name
F-box protein 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012170.3 216 Missense Mutation CAG,CGG Q64R NP_036302.1
NM_147188.2 216 Missense Mutation CAG,CGG Q64R NP_671717.1
XM_011521447.2 216 UTR 5 XP_011519749.1
Gene
UBE2Q2
Gene Name
ubiquitin conjugating enzyme E2 Q2
There are no transcripts associated with this gene.

View Full Product Details