Product Details

SNP ID
rs201241267
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:55418467 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATACTCCAAAGGAGCAAGTATTTT[C/A]AAGCCATATCAGAGCAACTGGATTT
Phenotype
MIM: 611326 MIM: 608706
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
C15orf65 PubMed Links

Gene Details

Gene
C15orf65
Gene Name
chromosome 15 open reading frame 65
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198784.1 507 Nonsense Mutation TAA,TCA *88S NP_001185713.1
Gene
CCPG1
Gene Name
cell cycle progression 1
There are no transcripts associated with this gene.

Gene
DYX1C1
Gene Name
dyslexia susceptibility 1 candidate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033559.2 507 Intron NP_001028731.1
NM_001033560.1 507 Intron NP_001028732.1
NM_130810.3 507 Intron NP_570722.2
Gene
DYX1C1-CCPG1
Gene Name
DYX1C1-CCPG1 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details