Product Details

SNP ID
rs201840540
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:45253467 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAGCCTGAGGGAAGCAAGAGGAC[C/T]GACGCACAAGGACACAGCCTGGGGG
Phenotype
MIM: 606208
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
LOC101928414 PubMed Links

Gene Details

Gene
LOC101928414
Gene Name
uncharacterized LOC101928414
There are no transcripts associated with this gene.

Gene
SLC28A2
Gene Name
solute carrier family 28 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004212.3 153 Silent Mutation ACC,ACT T39T NP_004203.2
XM_011522198.2 153 Silent Mutation ACC,ACT T39T XP_011520500.1
XM_011522199.1 153 Silent Mutation ACC,ACT T39T XP_011520501.1
XM_011522200.2 153 Silent Mutation ACC,ACT T39T XP_011520502.1
XM_011522201.2 153 Silent Mutation ACC,ACT T39T XP_011520503.1
XM_011522202.2 153 Intron XP_011520504.1

View Full Product Details