Product Details

SNP ID
rs201880670
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:41562581 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTCACAGTGGAGCCAAAAGATC[G/T]GGCAGTGCCACCCAATGCCCCTTTC
Phenotype
MIM: 600341
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TYRO3 PubMed Links

Gene Details

Gene
TYRO3
Gene Name
TYRO3 protein tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006293.3 480 Missense Mutation CGG,CTG R148L NP_006284.2
XM_017022542.1 480 Missense Mutation CGG,CTG R103L XP_016878031.1
XM_017022543.1 480 Missense Mutation CGG,CTG R148L XP_016878032.1

View Full Product Details