Product Details
- SNP ID
-
rs201617577
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:74956254 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGTCTGACCCTGCGTGGGCCCAGG[A/C]GGGAGGCTCTGCCACACCTCGCGTA
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RPP25
PubMed Links
Gene Details
- Gene
- RPP25
- Gene Name
- ribonuclease P/MRP subunit p25
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017793.2 |
1181 |
Silent Mutation |
CCG,CCT |
P110P |
NP_060263.2 |
View Full Product Details