Product Details
- SNP ID
-
rs201656241
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:58887061 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAGCGCTGCTACGATTCCCAGGGG[C/G]AGCGCCTCTCACACTGTGCCCTGAT
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SLTM
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2124203] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SLTM
- Gene Name
- SAFB like transcription modulator
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001013843.2 |
2656 |
Missense Mutation |
CCC,GCC |
P899A |
NP_001013865.1 |
NM_024755.3 |
2656 |
Missense Mutation |
CCC,GCC |
P917A |
NP_079031.2 |
XM_006720686.3 |
2656 |
Missense Mutation |
CCC,GCC |
P970A |
XP_006720749.3 |
XM_006720690.1 |
2656 |
Missense Mutation |
CCC,GCC |
P486A |
XP_006720753.1 |
XM_011522022.1 |
2656 |
Missense Mutation |
CCC,GCC |
P1059A |
XP_011520324.1 |
XM_011522023.1 |
2656 |
Missense Mutation |
CCC,GCC |
P1041A |
XP_011520325.1 |
XM_011522024.1 |
2656 |
Missense Mutation |
CCC,GCC |
P1009A |
XP_011520326.1 |
XM_011522025.1 |
2656 |
Missense Mutation |
CCC,GCC |
P993A |
XP_011520327.1 |
XM_011522026.2 |
2656 |
Missense Mutation |
CCC,GCC |
P986A |
XP_011520328.2 |
XM_011522027.1 |
2656 |
Missense Mutation |
CCC,GCC |
P977A |
XP_011520329.1 |
XM_011522028.1 |
2656 |
Missense Mutation |
CCC,GCC |
P959A |
XP_011520330.1 |
XM_011522029.2 |
2656 |
Missense Mutation |
CCC,GCC |
P871A |
XP_011520331.1 |
XM_011522030.2 |
2656 |
Intron |
|
|
XP_011520332.1 |
XM_011522031.2 |
2656 |
Missense Mutation |
CCC,GCC |
P689A |
XP_011520333.1 |
XM_017022576.1 |
2656 |
Missense Mutation |
CCC,GCC |
P849A |
XP_016878065.1 |
XM_017022577.1 |
2656 |
Missense Mutation |
CCC,GCC |
P968A |
XP_016878066.1 |
XM_017022578.1 |
2656 |
Missense Mutation |
CCC,GCC |
P952A |
XP_016878067.1 |
XM_017022579.1 |
2656 |
Missense Mutation |
CCC,GCC |
P864A |
XP_016878068.1 |
XM_017022580.1 |
2656 |
Missense Mutation |
CCC,GCC |
P486A |
XP_016878069.1 |
View Full Product Details