Product Details

SNP ID
rs201883358
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:88468013 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAACAGAACGCGGCTCCCAGCCA[C/T]ACCAAGTTCAGGTGAGCCCCACTTG
Phenotype
MIM: 611851 MIM: 611977
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRPL46 PubMed Links

Gene Details

Gene
MRPL46
Gene Name
mitochondrial ribosomal protein L46
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022163.3 433 Intron NP_071446.2
Gene
MRPS11
Gene Name
mitochondrial ribosomal protein S11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321970.1 433 Silent Mutation CAC,CAT H56H NP_001308899.1
NM_001321972.1 433 UTR 5 NP_001308901.1
NM_001321973.1 433 Silent Mutation CAC,CAT H56H NP_001308902.1
NM_001321974.1 433 Silent Mutation CAC,CAT H57H NP_001308903.1
NM_001321976.1 433 UTR 5 NP_001308905.1
NM_022839.4 433 Silent Mutation CAC,CAT H57H NP_073750.2
NM_176805.3 433 Silent Mutation CAC,CAT H57H NP_789775.1

View Full Product Details