Product Details

SNP ID
rs201738101
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:62067918 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCACCTGCCCCGTGTGCGCACCG[C/T]CTACGGCTTCTGCGCGCTGCTCGAG
Phenotype
MIM: 610343 MIM: 608879
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C2CD4A PubMed Links

Gene Details

Gene
C2CD4A
Gene Name
C2 calcium dependent domain containing 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207322.2 446 Missense Mutation GCC,GTC A102V NP_997205.2
Gene
LOC101928907
Gene Name
uncharacterized LOC101928907
There are no transcripts associated with this gene.

Gene
VPS13C
Gene Name
vacuolar protein sorting 13 homolog C
There are no transcripts associated with this gene.

View Full Product Details