Product Details
- SNP ID
-
rs200499926
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:99138119 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCCTGCCTTGGCTTCGAAGCAACC[C/T]TGGGGGCCCTGCAGACAAGCAGAGG
- Phenotype
-
MIM: 606087
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SYNM
PubMed Links
Gene Details
- Gene
- SYNM
- Gene Name
- synemin
There are no transcripts associated with this gene.
- Gene
- TTC23
- Gene Name
- tetratricopeptide repeat domain 23
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001288615.1 |
1627 |
Missense Mutation |
AAG,AGG |
K412R |
NP_001275544.1 |
NM_001288616.1 |
1627 |
Missense Mutation |
AAG,AGG |
K412R |
NP_001275545.1 |
XM_011521927.2 |
1627 |
Silent Mutation |
CAA,CAG |
Q484Q |
XP_011520229.1 |
XM_011521929.1 |
1627 |
Silent Mutation |
CAA,CAG |
Q487Q |
XP_011520231.1 |
XM_011521930.1 |
1627 |
Silent Mutation |
CAA,CAG |
Q487Q |
XP_011520232.1 |
XM_011521931.1 |
1627 |
Silent Mutation |
CAA,CAG |
Q487Q |
XP_011520233.1 |
XM_011521932.1 |
1627 |
Silent Mutation |
CAA,CAG |
Q487Q |
XP_011520234.1 |
XM_011521933.1 |
1627 |
Silent Mutation |
CAA,CAG |
Q487Q |
XP_011520235.1 |
XM_011521934.1 |
1627 |
Silent Mutation |
CAA,CAG |
Q487Q |
XP_011520236.1 |
XM_011521935.1 |
1627 |
Silent Mutation |
CAA,CAG |
Q487Q |
XP_011520237.1 |
XM_011521936.1 |
1627 |
Silent Mutation |
CAA,CAG |
Q487Q |
XP_011520238.1 |
XM_011521937.1 |
1627 |
Silent Mutation |
CAA,CAG |
Q487Q |
XP_011520239.1 |
XM_011521939.1 |
1627 |
Silent Mutation |
CAA,CAG |
Q487Q |
XP_011520241.1 |
XM_011521940.2 |
1627 |
Silent Mutation |
CAA,CAG |
Q459Q |
XP_011520242.1 |
XM_011521941.2 |
1627 |
Missense Mutation |
AGG,GGG |
R437G |
XP_011520243.1 |
XM_011521943.2 |
1627 |
Intron |
|
|
XP_011520245.1 |
XM_011521944.2 |
1627 |
Intron |
|
|
XP_011520246.1 |
XM_011521945.2 |
1627 |
Intron |
|
|
XP_011520247.1 |
XM_017022509.1 |
1627 |
Silent Mutation |
CAA,CAG |
Q487Q |
XP_016877998.1 |
XM_017022510.1 |
1627 |
Silent Mutation |
CAA,CAG |
Q487Q |
XP_016877999.1 |
XM_017022511.1 |
1627 |
Silent Mutation |
CAA,CAG |
Q487Q |
XP_016878000.1 |
XM_017022512.1 |
1627 |
Missense Mutation |
AAG,AGG |
K412R |
XP_016878001.1 |
XM_017022513.1 |
1627 |
Missense Mutation |
AAG,AGG |
K412R |
XP_016878002.1 |
XM_017022514.1 |
1627 |
Missense Mutation |
AAG,AGG |
K412R |
XP_016878003.1 |
XM_017022515.1 |
1627 |
Missense Mutation |
AAG,AGG |
K412R |
XP_016878004.1 |
XM_017022516.1 |
1627 |
Missense Mutation |
AAG,AGG |
K412R |
XP_016878005.1 |
XM_017022517.1 |
1627 |
Missense Mutation |
AAG,AGG |
K412R |
XP_016878006.1 |
XM_017022518.1 |
1627 |
Missense Mutation |
AAG,AGG |
K412R |
XP_016878007.1 |
View Full Product Details