Product Details

SNP ID
rs200789139
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:74749857 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCTCGGGTCCCCAAAGGCCTGA[A/G]AAGTCCACCAGAGCCATGGGGCTGG
Phenotype
MIM: 124060
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYP1A2 PubMed Links

Gene Details

Gene
CYP1A2
Gene Name
cytochrome P450 family 1 subfamily A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000761.4 183 Missense Mutation AAA,AGA K40R NP_000752.2

View Full Product Details