Product Details

SNP ID
rs199841831
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:48121148 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCAGGGACCTCCCTGCCCCAAC[A/G]TCTCCCAAGGGCCACAGGTAGGTGG
Phenotype
MIM: 609802
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC24A5 PubMed Links

Gene Details

Gene
SLC24A5
Gene Name
solute carrier family 24 member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_205850.2 177 Missense Mutation CAT,CGT H35R NP_995322.1
XM_017022079.1 177 Intron XP_016877568.1
XM_017022080.1 177 Intron XP_016877569.1

View Full Product Details