Product Details

SNP ID
rs201109162
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40807645 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCCGCTGCCGTACGCTGGCTGCA[C/G]GAGAGCGTCCGGATTCCCTGCTCTA
Phenotype
MIM: 616844 MIM: 613256
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC17 PubMed Links

Gene Details

Gene
DNAJC17
Gene Name
DnaJ heat shock protein family (Hsp40) member C17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018163.2 570 Intron NP_060633.1
XM_017022401.1 570 Intron XP_016877890.1
XM_017022402.1 570 Intron XP_016877891.1
Gene
PPP1R14D
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 14D
There are no transcripts associated with this gene.

Gene
ZFYVE19
Gene Name
zinc finger FYVE-type containing 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077268.1 570 Missense Mutation ACG,AGG T19R NP_001070736.1
NM_001258420.1 570 Missense Mutation ACG,AGG T19R NP_001245349.1
NM_001258421.1 570 UTR 5 NP_001245350.1
NM_032850.4 570 Intron NP_116239.3
XM_017022683.1 570 Intron XP_016878172.1
XM_017022684.1 570 Missense Mutation ACG,AGG T19R XP_016878173.1

View Full Product Details