Product Details

SNP ID
rs199517293
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:66551803 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCTGAGTATCCTTTCTCCCATTC[A/G]AACTTATCCAACAGAGACCAGGAAG
Phenotype
MIM: 609984
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LCTL PubMed Links

Gene Details

Gene
LCTL
Gene Name
lactase like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278562.1 1515 Silent Mutation TTC,TTT F288F NP_001265491.1
NM_207338.3 1515 Silent Mutation TTC,TTT F461F NP_997221.2
XM_017021999.1 1515 Silent Mutation TTC,TTT F478F XP_016877488.1
Gene
ZWILCH
Gene Name
zwilch kinetochore protein
There are no transcripts associated with this gene.

View Full Product Details