Product Details

SNP ID
rs200508337
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78098051 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTGCACCATTACCAGGAGGCAC[A/G]GCTCGAGCCCTTCAAAGAGATGCTG
Phenotype
MIM: 605564
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CIB2 PubMed Links

Gene Details

Gene
CIB2
Gene Name
calcium and integrin binding family member 2
There are no transcripts associated with this gene.

Gene
SH2D7
Gene Name
SH2 domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101404.1 389 Missense Mutation CAG,CGG Q130R NP_001094874.1
XM_011521895.2 389 Missense Mutation CAG,CGG Q130R XP_011520197.1
XM_011521896.2 389 Missense Mutation CAG,CGG Q97R XP_011520198.1
XM_011521899.2 389 Missense Mutation CAG,CGG Q58R XP_011520201.1
XM_017022487.1 389 Missense Mutation CAG,CGG Q111R XP_016877976.1
XM_017022488.1 389 UTR 5 XP_016877977.1

View Full Product Details