Product Details

SNP ID
rs200763849
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78513241 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGATGACCAAAACTTTCATGTCTA[C/T]GTTTCAAAAACCAAAGATGGCCCAA
Phenotype
MIM: 614681
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HYKK PubMed Links

Gene Details

Gene
HYKK
Gene Name
hydroxylysine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013619.3 266 Silent Mutation TAC,TAT Y51Y NP_001013641.2
NM_001083612.1 266 Silent Mutation TAC,TAT Y51Y NP_001077081.1
XM_011521231.2 266 Silent Mutation TAC,TAT Y51Y XP_011519533.1
XM_017021922.1 266 Silent Mutation TAC,TAT Y51Y XP_016877411.1

View Full Product Details