Product Details

SNP ID
rs199595126
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:90904312 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGAGTGCAACACGATCCCACCCG[A/C]CACCAGAATGGTGGGAACTTCCCCC
Phenotype
MIM: 190030 MIM: 600988
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FES PubMed Links

Gene Details

Gene
FES
Gene Name
FES proto-oncogene, tyrosine kinase
There are no transcripts associated with this gene.

Gene
MAN2A2
Gene Name
mannosidase alpha class 2A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320977.1 273 Silent Mutation CGA,CGC R35R NP_001307906.1
NM_006122.3 273 Silent Mutation CGA,CGC R35R NP_006113.2
XM_005254910.3 273 Silent Mutation CGA,CGC R35R XP_005254967.1
XM_011521565.2 273 Intron XP_011519867.1
XM_011521566.2 273 Intron XP_011519868.1
XM_017022184.1 273 Silent Mutation CGA,CGC R35R XP_016877673.1
XM_017022185.1 273 Silent Mutation CGA,CGC R35R XP_016877674.1

View Full Product Details