Product Details

SNP ID
rs200664200
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:34752898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGAGCTTGGATTTTGATGCAGT[A/G]CGTAGACCTGATGGGTGTGGAGTCA
Phenotype
MIM: 607058
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GJD2 PubMed Links

Gene Details

Gene
GJD2
Gene Name
gap junction protein delta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020660.2 639 Silent Mutation CGC,CGT R182R NP_065711.1
XM_017022438.1 639 Silent Mutation CGC,CGT R131R XP_016877927.1
Gene
LOC101928174
Gene Name
uncharacterized LOC101928174
There are no transcripts associated with this gene.

View Full Product Details