Product Details

SNP ID
rs200775057
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40569679 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGGAGCGTGTCGTGCCGCCCCC[A/G]AAGAAGCGGCGGACCGGGGTGAGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C15orf57 PubMed Links

Gene Details

Gene
C15orf57
Gene Name
chromosome 15 open reading frame 57
There are no transcripts associated with this gene.

Gene
RPUSD2
Gene Name
RNA pseudouridylate synthase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286407.1 387 Silent Mutation CCA,CCG P114P NP_001273336.1
NM_152260.2 387 Silent Mutation CCA,CCG P114P NP_689473.1

View Full Product Details