Product Details

SNP ID
rs201080470
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:48826055 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACTTCGCTTCCAGAGGAGATGAT[G/T]GGCAAACTGTTATCCATATGGTATC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SHC4 PubMed Links

Gene Details

Gene
SHC4
Gene Name
SHC adaptor protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203349.3 2238 Silent Mutation CCA,CCC P603P NP_976224.3
XM_005254375.3 2238 Silent Mutation CCA,CCC P420P XP_005254432.1

View Full Product Details