Product Details
- SNP ID
-
rs199572545
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:100402756 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTGGGAATGAGGTGCCTCTCAGCC[A/C]TGAGGCCGTTCTTTAAACAATCCAT
- Phenotype
-
MIM: 615276
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CERS3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2439928] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CERS3
- Gene Name
- ceramide synthase 3
- Gene
- CERS3-AS1
- Gene Name
- CERS3 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details