Product Details
- SNP ID
-
rs199717400
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:83107723 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACCGGGGTCTTCGTGCTGTCCCTC[C/T]CGGCCATCCCGGTCACCTATGTCTT
- Phenotype
-
MIM: 616643
MIM: 606562
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HDGFRP3
PubMed Links
Gene Details
- Gene
- HDGFRP3
- Gene Name
- hepatoma-derived growth factor, related protein 3
There are no transcripts associated with this gene.
- Gene
- LOC105370931
- Gene Name
- uncharacterized LOC105370931
There are no transcripts associated with this gene.
- Gene
- TM6SF1
- Gene Name
- transmembrane 6 superfamily member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001144903.1 |
90 |
Missense Mutation |
CCG,TCG |
P15S |
NP_001138375.1 |
NM_023003.3 |
90 |
Missense Mutation |
CCG,TCG |
P15S |
NP_075379.2 |
XM_011521679.2 |
90 |
Missense Mutation |
CCG,TCG |
P15S |
XP_011519981.1 |
XM_011521680.2 |
90 |
Missense Mutation |
CCG,TCG |
P15S |
XP_011519982.1 |
XM_017022320.1 |
90 |
Missense Mutation |
CCG,TCG |
P15S |
XP_016877809.1 |
XM_017022321.1 |
90 |
Missense Mutation |
CCG,TCG |
P15S |
XP_016877810.1 |
XM_017022322.1 |
90 |
Missense Mutation |
CCG,TCG |
P15S |
XP_016877811.1 |
XM_017022323.1 |
90 |
UTR 5 |
|
|
XP_016877812.1 |
XM_017022324.1 |
90 |
UTR 5 |
|
|
XP_016877813.1 |
XM_017022325.1 |
90 |
UTR 5 |
|
|
XP_016877814.1 |
XM_017022326.1 |
90 |
UTR 5 |
|
|
XP_016877815.1 |
XM_017022327.1 |
90 |
UTR 5 |
|
|
XP_016877816.1 |
XM_017022328.1 |
90 |
Missense Mutation |
CCG,TCG |
P15S |
XP_016877817.1 |
XM_017022329.1 |
90 |
Missense Mutation |
CCG,TCG |
P15S |
XP_016877818.1 |
XM_017022330.1 |
90 |
Missense Mutation |
CCG,TCG |
P15S |
XP_016877819.1 |
XM_017022331.1 |
90 |
Missense Mutation |
CCG,TCG |
P15S |
XP_016877820.1 |
View Full Product Details