Product Details

SNP ID
rs200229830
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40815707 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGTCTCTCAGGCTTATTTCTGAG[C/G]CCGGCTGAGTCTCCGGAGTTTCTTG
Phenotype
MIM: 616844 MIM: 613256
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC17 PubMed Links

Gene Details

Gene
DNAJC17
Gene Name
DnaJ heat shock protein family (Hsp40) member C17
There are no transcripts associated with this gene.

Gene
PPP1R14D
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 14D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130143.1 607 Silent Mutation GGC,GGG G181G NP_001123615.1
NM_017726.7 607 Missense Mutation CCT,GCT P143A NP_060196.1
XM_017022372.1 607 UTR 3 XP_016877861.1
Gene
ZFYVE19
Gene Name
zinc finger FYVE-type containing 19
There are no transcripts associated with this gene.

View Full Product Details