Product Details
- SNP ID
-
rs200633089
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:77614647 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTCCCGGATGCGGGCGCGGCGGCA[A/G]GTGAAGTAGTTGGGCAGTAGCACAT
- Phenotype
-
MIM: 609791
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LINGO1
PubMed Links
Gene Details
- Gene
- LINGO1
- Gene Name
- leucine rich repeat and Ig domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001301186.1 |
1454 |
Silent Mutation |
ACC,ACT |
T414T |
NP_001288115.1 |
NM_001301187.1 |
1454 |
Silent Mutation |
ACC,ACT |
T414T |
NP_001288116.1 |
NM_001301189.1 |
1454 |
Silent Mutation |
ACC,ACT |
T414T |
NP_001288118.1 |
NM_001301191.1 |
1454 |
Silent Mutation |
ACC,ACT |
T414T |
NP_001288120.1 |
NM_001301192.1 |
1454 |
Silent Mutation |
ACC,ACT |
T414T |
NP_001288121.1 |
NM_001301194.1 |
1454 |
Silent Mutation |
ACC,ACT |
T414T |
NP_001288123.1 |
NM_001301195.1 |
1454 |
Silent Mutation |
ACC,ACT |
T414T |
NP_001288124.1 |
NM_001301197.1 |
1454 |
Silent Mutation |
ACC,ACT |
T414T |
NP_001288126.1 |
NM_001301198.1 |
1454 |
Silent Mutation |
ACC,ACT |
T414T |
NP_001288127.1 |
NM_001301199.1 |
1454 |
Silent Mutation |
ACC,ACT |
T414T |
NP_001288128.1 |
NM_001301200.1 |
1454 |
Silent Mutation |
ACC,ACT |
T414T |
NP_001288129.1 |
NM_032808.6 |
1454 |
Silent Mutation |
ACC,ACT |
T420T |
NP_116197.4 |
XM_011522118.2 |
1454 |
Silent Mutation |
ACC,ACT |
T414T |
XP_011520420.1 |
XM_017022682.1 |
1454 |
Silent Mutation |
ACC,ACT |
T414T |
XP_016878171.1 |
View Full Product Details