Product Details

SNP ID
rs200157375
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:74839641 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAGTCCTGGAAGGAGATGCGAC[A/G]GCTGGGGTCCCGCTCCAGGAGCCGC
Phenotype
MIM: 609585 MIM: 606912 MIM: 613472
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CPLX3 PubMed Links

Gene Details

Gene
CPLX3
Gene Name
complexin 3
There are no transcripts associated with this gene.

Gene
MIR6882
Gene Name
microRNA 6882
There are no transcripts associated with this gene.

Gene
SCAMP2
Gene Name
secretory carrier membrane protein 2
There are no transcripts associated with this gene.

Gene
ULK3
Gene Name
unc-51 like kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099436.3 928 Missense Mutation CGT,TGT R257C NP_001092906.3
NM_001284364.2 928 Missense Mutation CGT,TGT R257C NP_001271293.2
NM_001284365.2 928 Missense Mutation CGT,TGT R140C NP_001271294.1
XM_005254289.2 928 Missense Mutation CGT,TGT R268C XP_005254346.1
XM_017022068.1 928 Missense Mutation CGT,TGT R268C XP_016877557.1
XM_017022069.1 928 Missense Mutation CGT,TGT R167C XP_016877558.1
XM_017022070.1 928 Missense Mutation CGT,TGT R167C XP_016877559.1
XM_017022071.1 928 Missense Mutation CGT,TGT R167C XP_016877560.1
XM_017022072.1 928 Missense Mutation CGT,TGT R167C XP_016877561.1

View Full Product Details