Product Details

SNP ID
rs201104442
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43805103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAAGCAGTTGGACCCTAGGTAGT[C/T]TTCCGCTTCTTGGCAGATGGCCGCT
Phenotype
MIM: 612784 MIM: 600215 MIM: 605054 MIM: 614550
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HYPK PubMed Links

Gene Details

Gene
HYPK
Gene Name
huntingtin interacting protein K
There are no transcripts associated with this gene.

Gene
MFAP1
Gene Name
microfibrillar associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005926.2 1495 Silent Mutation AAA,AAG K437K NP_005917.2
Gene
SERF2
Gene Name
small EDRK-rich factor 2
There are no transcripts associated with this gene.

Gene
SERF2-C15ORF63
Gene Name
SERF2-C15orf63 readthrough
There are no transcripts associated with this gene.

Gene
SERINC4
Gene Name
serine incorporator 4
There are no transcripts associated with this gene.

View Full Product Details