Product Details

SNP ID
rs199864572
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:88467724 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGGGTCAATTCAAGTCATGCAG[A/G]CTGTGAGAAACGCGGGGTCGCGGTT
Phenotype
MIM: 611851 MIM: 611977
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPL46 PubMed Links

Gene Details

Gene
MRPL46
Gene Name
mitochondrial ribosomal protein L46
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022163.3 272 Intron NP_071446.2
Gene
MRPS11
Gene Name
mitochondrial ribosomal protein S11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321970.1 272 Missense Mutation ACT,GCT T3A NP_001308899.1
NM_001321972.1 272 UTR 5 NP_001308901.1
NM_001321973.1 272 Missense Mutation ACT,GCT T3A NP_001308902.1
NM_001321974.1 272 Missense Mutation ACT,GCT T3A NP_001308903.1
NM_001321976.1 272 UTR 5 NP_001308905.1
NM_022839.4 272 Missense Mutation ACT,GCT T3A NP_073750.2
NM_176805.3 272 Missense Mutation ACT,GCT T3A NP_789775.1

View Full Product Details