Product Details

SNP ID
rs200308396
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:70832715 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCTGTGATCATGAACTCATGGGC[A/T]TTGATGGCTGCTTCCACCTCCTCGA
Phenotype
MIM: 611300
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LARP6 PubMed Links

Gene Details

Gene
LARP6
Gene Name
La ribonucleoprotein domain family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286679.1 725 Missense Mutation AAA,AAT K87N NP_001273608.1
NM_018357.3 725 Missense Mutation AAA,AAT K271N NP_060827.2
NM_197958.2 725 Intron NP_932062.1

View Full Product Details