Product Details

SNP ID
rs199760617
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:73740188 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTCCACCTCCTGCTCACCGTCC[C/T]CCTCCTCATCGTCACAGCACAGAGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C15orf59 PubMed Links

Gene Details

Gene
C15orf59
Gene Name
chromosome 15 open reading frame 59
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039614.2 839 Missense Mutation GAG,GGG E204G NP_001034703.1
NM_001303254.1 839 Missense Mutation GAG,GGG E176G NP_001290183.1
XM_005254369.4 839 Missense Mutation GAG,GGG E204G XP_005254426.1

View Full Product Details