Product Details

SNP ID
rs201135688
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78513293 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAATATGTCCTCAAAATAAGCAAC[A/G]CCAAGGCTAGCAAAAATCCAGACCT
Phenotype
MIM: 614681
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HYKK PubMed Links

Gene Details

Gene
HYKK
Gene Name
hydroxylysine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013619.3 318 Missense Mutation ACC,GCC T69A NP_001013641.2
NM_001083612.1 318 Missense Mutation ACC,GCC T69A NP_001077081.1
XM_011521231.2 318 Missense Mutation ACC,GCC T69A XP_011519533.1
XM_017021922.1 318 Missense Mutation ACC,GCC T69A XP_016877411.1

View Full Product Details