Product Details

SNP ID
rs199507109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:74410909 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGTGGCGCCATGAGTAGGTGGC[A/G]TGGCGGGATTCCATGGGGCAGCTCA
Phenotype
MIM: 607961
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR6881 PubMed Links

Gene Details

Gene
MIR6881
Gene Name
microRNA 6881
There are no transcripts associated with this gene.

Gene
SEMA7A
Gene Name
semaphorin 7A (John Milton Hagen blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146029.2 1653 Silent Mutation CAC,CAT H558H NP_001139501.1
NM_001146030.2 1653 Silent Mutation CAC,CAT H407H NP_001139502.1
NM_003612.4 1653 Silent Mutation CAC,CAT H572H NP_003603.1

View Full Product Details