Product Details

SNP ID
rs199512646
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2682737 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCACCACTCGGCAGACCCTGTG[C/T]CGGGACCCGAAATCCTTCCTGTACC
Phenotype
MIM: 611285
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ERVK13-1 PubMed Links

Gene Details

Gene
ERVK13-1
Gene Name
endogenous retrovirus group K13 member 1
There are no transcripts associated with this gene.

Gene
KCTD5
Gene Name
potassium channel tetramerization domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018992.3 244 Silent Mutation TGC,TGT C63C NP_061865.1

View Full Product Details