Product Details
- SNP ID
-
rs199541169
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:1985946 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCCACCTGCACAATGAAGTGAACC[A/G]CAAGCTGGGCAAGCCTGACTTCGAC
- Phenotype
-
MIM: 600924
MIM: 611256
MIM: 603927
MIM: 605915
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GFER
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs36041021] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GFER
- Gene Name
- growth factor, augmenter of liver regeneration
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005262.2 |
606 |
Missense Mutation |
CAC,CGC |
H179R |
NP_005253.3 |
- Gene
- NOXO1
- Gene Name
- NADPH oxidase organizer 1
There are no transcripts associated with this gene.
- Gene
- SYNGR3
- Gene Name
- synaptogyrin 3
There are no transcripts associated with this gene.
- Gene
- TBL3
- Gene Name
- transducin beta like 3
There are no transcripts associated with this gene.
View Full Product Details