Product Details

SNP ID
rs200427925
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30748841 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGATGACGCTGGACGTGGGGCC[A/G]GAGGATGAGCTGCCCGACTGGGCCG
Phenotype
MIM: 172471 MIM: 611421
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CCDC189 PubMed Links

Gene Details

Gene
CCDC189
Gene Name
coiled-coil domain containing 189
There are no transcripts associated with this gene.

Gene
PHKG2
Gene Name
phosphorylase kinase catalytic subunit gamma 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000294.2 231 Silent Mutation CCA,CCG P7P NP_000285.1
NM_001172432.1 231 Silent Mutation CCA,CCG P7P NP_001165903.1
Gene
SRCAP
Gene Name
Snf2-related CREBBP activator protein
There are no transcripts associated with this gene.

Gene
TMEM265
Gene Name
transmembrane protein 265
There are no transcripts associated with this gene.

View Full Product Details