Product Details

SNP ID
rs201080187
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29808686 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGAGCTCGGCTTATATTTCGGA[C/G]CACATGAAGGTGCACAGCCAGGGTC
Phenotype
MIM: 603213 MIM: 600999 MIM: 612033 MIM: 614386
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KIF22 PubMed Links

Gene Details

Gene
KIF22
Gene Name
kinesin family member 22
There are no transcripts associated with this gene.

Gene
MAZ
Gene Name
MYC associated zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042539.2 1204 Missense Mutation GAC,GAG D408E NP_001036004.1
NM_001276275.1 1204 Missense Mutation GAC,GAG D385E NP_001263204.1
NM_001276276.1 1204 Missense Mutation GAC,GAG D103E NP_001263205.1
NM_002383.3 1204 Missense Mutation GAC,GAG D408E NP_002374.2
XM_006721047.3 1204 Missense Mutation GAC,GAG D385E XP_006721110.1
XM_006721048.3 1204 Intron XP_006721111.1
Gene
PAGR1
Gene Name
PAXIP1 associated glutamate rich protein 1
There are no transcripts associated with this gene.

Gene
PRRT2
Gene Name
proline rich transmembrane protein 2
There are no transcripts associated with this gene.

View Full Product Details