Product Details

SNP ID
rs201112184
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1985945 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCCACCTGCACAATGAAGTGAAC[C/T]GCAAGCTGGGCAAGCCTGACTTCGA
Phenotype
MIM: 600924 MIM: 611256 MIM: 603927 MIM: 605915
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GFER PubMed Links

Gene Details

Gene
GFER
Gene Name
growth factor, augmenter of liver regeneration
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005262.2 605 Missense Mutation CGC,TGC R179C NP_005253.3
Gene
NOXO1
Gene Name
NADPH oxidase organizer 1
There are no transcripts associated with this gene.

Gene
SYNGR3
Gene Name
synaptogyrin 3
There are no transcripts associated with this gene.

Gene
TBL3
Gene Name
transducin beta like 3
There are no transcripts associated with this gene.

View Full Product Details