Product Details

SNP ID
rs200201528
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:20478576 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCCAATCTGCTTCTTTCTCCAG[A/C]TCATAGATGATAAGGGCAACGTCCT
Phenotype
MIM: 614358
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ACSM2A PubMed Links

Gene Details

Gene
ACSM2A
Gene Name
acyl-CoA synthetase medium-chain family member 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308169.1 Intron NP_001295098.1
NM_001308172.1 Intron NP_001295101.1
NM_001308954.1 Intron NP_001295883.1
XM_017022923.1 Intron XP_016878412.1
XM_017022924.1 Intron XP_016878413.1
XM_017022925.1 Intron XP_016878414.1
XM_017022926.1 Intron XP_016878415.1

View Full Product Details