Product Details

SNP ID
rs200824527
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2471917 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGGGCCTGCGACGCCTCCGACCC[G/T]CGACGGGCACACTCCCCCGCCCTCC
Phenotype
MIM: 602349 MIM: 613577
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C16orf59 PubMed Links

Gene Details

Gene
C16orf59
Gene Name
chromosome 16 open reading frame 59
There are no transcripts associated with this gene.

Gene
LOC729652
Gene Name
uncharacterized LOC729652
There are no transcripts associated with this gene.

Gene
MIR6768
Gene Name
microRNA 6768
There are no transcripts associated with this gene.

Gene
NTN3
Gene Name
netrin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006181.2 419 Silent Mutation CCG,CCT P72P NP_006172.1
Gene
TBC1D24
Gene Name
TBC1 domain family member 24
There are no transcripts associated with this gene.

View Full Product Details