Product Details

SNP ID
rs201342647
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:48083933 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCCCATTTTCCATAACCAGGAC[A/G]TGATCGCAGTTGAGAACTGTGTTGA
Phenotype
MIM: 607041
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCC12 PubMed Links

Gene Details

Gene
ABCC12
Gene Name
ATP binding cassette subfamily C member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033226.2 4315 Silent Mutation CAC,CAT H1323H NP_150229.2

View Full Product Details