Product Details

SNP ID
rs200433383
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:22347814 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCTCAGCGAACACATGATCATA[A/C]ACGAAGTGTCTAGGGAAAAAAATAT
Phenotype
MIM: 117340
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
CDR2 PubMed Links

Gene Details

Gene
CDR2
Gene Name
cerebellar degeneration related protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001802.1 824 Silent Mutation GTG,GTT V172V NP_001793.1

View Full Product Details