Product Details

SNP ID
rs200826725
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30985753 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGTGGTGCGAATGCTGCTGCAGC[A/G]GGAGCCCCGGCTCGGGGAGCTGCGG
Phenotype
MIM: 607764 MIM: 611052 MIM: 601485
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSD3B7 PubMed Links

Gene Details

Gene
HSD3B7
Gene Name
hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142777.1 201 Missense Mutation CAG,CGG Q32R NP_001136249.1
NM_001142778.1 201 Missense Mutation CAG,CGG Q32R NP_001136250.1
NM_025193.3 201 Missense Mutation CAG,CGG Q32R NP_079469.2
XM_005255601.3 201 Missense Mutation CAG,CGG Q32R XP_005255658.2
XM_011545960.2 201 Missense Mutation CAG,CGG Q32R XP_011544262.1
XM_011545961.1 201 Missense Mutation CAG,CGG Q32R XP_011544263.1
XM_011545962.2 201 Missense Mutation CAG,CGG Q32R XP_011544264.1
XM_017023732.1 201 Missense Mutation CAG,CGG Q32R XP_016879221.1
Gene
SETD1A
Gene Name
SET domain containing 1A
There are no transcripts associated with this gene.

Gene
STX1B
Gene Name
syntaxin 1B
There are no transcripts associated with this gene.

View Full Product Details