Product Details

SNP ID
rs201087605
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1999659 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCATGTGTCCATTAACGACGAC[A/G]GCAACTGGCCCCTCGGCTCTGCTGG
Phenotype
MIM: 603927
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SYNGR3 PubMed Links

Gene Details

Gene
SYNGR3
Gene Name
synaptogyrin 3
There are no transcripts associated with this gene.

Gene
ZNF598
Gene Name
zinc finger protein 598
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178167.3 1964 Silent Mutation GCC,GCT A630A NP_835461.2

View Full Product Details