Product Details

SNP ID
rs201280263
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29968182 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGGCTGGAAATGCCACCCTGAG[C/T]CCTAGAATGGGTGAGGAATGTGTTA
Phenotype
MIM: 613199
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC107987231 PubMed Links

Gene Details

Gene
LOC107987231
Gene Name
uncharacterized LOC107987231
There are no transcripts associated with this gene.

Gene
TAOK2
Gene Name
TAO kinase 2
There are no transcripts associated with this gene.

Gene
TMEM219
Gene Name
transmembrane protein 219
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083613.1 652 Silent Mutation AGC,AGT S171S NP_001077082.1
NM_194280.3 652 Silent Mutation AGC,AGT S171S NP_919256.1
XM_005255123.1 652 Silent Mutation AGC,AGT S171S XP_005255180.1

View Full Product Details