Product Details

SNP ID
rs201565784
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1979521 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTGCGCGGCTGCTCTCGTAGGCG[C/T]GGGAAGCACAGAACTGGGGACCTGG
Phenotype
MIM: 600924 MIM: 611256 MIM: 605915
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GFER PubMed Links

Gene Details

Gene
GFER
Gene Name
growth factor, augmenter of liver regeneration
There are no transcripts associated with this gene.

Gene
NOXO1
Gene Name
NADPH oxidase organizer 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267721.1 1104 Missense Mutation CAC,CGC H245R NP_001254650.1
NM_144603.3 1104 Missense Mutation CAC,CGC H240R NP_653204.1
NM_172167.2 1104 Missense Mutation CAC,CGC H241R NP_751907.1
NM_172168.2 1104 Missense Mutation CAC,CGC H246R NP_751908.1
XM_005255099.4 1104 Missense Mutation CAC,CGC H246R XP_005255156.1
XM_017022927.1 1104 Missense Mutation CAC,CGC H246R XP_016878416.1
XM_017022928.1 1104 Missense Mutation CAC,CGC H246R XP_016878417.1
Gene
TBL3
Gene Name
transducin beta like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006453.2 1104 Intron NP_006444.2

View Full Product Details