Product Details

SNP ID
rs199844241
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:19072014 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGGCAGCTGTGGATCGAATAATC[C/T]GGGTGGATCATGCAGGCGAATATGG
Phenotype
MIM: 601683
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COQ7 PubMed Links

Gene Details

Gene
COQ7
Gene Name
coenzyme Q7, hydroxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190983.1 179 Missense Mutation CGG,TGG R16W NP_001177912.1
NM_016138.4 179 Missense Mutation CGG,TGG R54W NP_057222.2
Gene
LOC102723385
Gene Name
uncharacterized LOC102723385
There are no transcripts associated with this gene.

Gene
TMC7
Gene Name
transmembrane channel like 7
There are no transcripts associated with this gene.

View Full Product Details